Assay Details
Gene Symbol: MPZL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015673.2 | NP_001015673.1 | 3 - 4 | 676 | 82 | |
GenBank mRNA | BC102486.1 | - | 3 - 4 | 568 | 82 | |
BT020659.1 | - | 3 - 4 | 677 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
