Assay Details
Gene Symbol: TADA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024511.1 | NP_001019682.1 | 4 - 5 | 655 | 86 | |
GenBank mRNA | BC110248.1 | - | 3 - 4 | 581 | 86 | |
BT020627.1 | - | 4 - 5 | 655 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
