Assay Details
Gene Symbol: HDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024551.1 | NP_001019722.1 | 1 - 2 | 51 | 66 | |
GenBank mRNA | BC114752.1 | - | 1 - 2 | 73 | 66 | |
BT020686.1 | - | 1 - 2 | 51 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
