Assay Details
Gene Symbol: IST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034335.2 | NP_001029507.1 | 2 - 3 | 151 | 87 | |
GenBank mRNA | BC103094.1 | - | 2 - 3 | 153 | 87 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC103094.1, NM_001034335.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
