Assay Details
Gene Symbol: IST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034335.2 | NP_001029507.1 | 9 - 10 | 1002 | 113 | |
GenBank mRNA | BC103094.1 | - | 9 - 10 | 1004 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
