Assay Details
Gene Symbol: TMEM14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034379.1 | NP_001029551.1 | 4 - 5 | 324 | 112 | |
GenBank mRNA | BC102864.1 | - | 4 - 5 | 324 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
