Assay Details
Gene Symbol: PA2G4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034699.1 | NP_001029871.1 | 2 - 3 | 372 | 159 | |
GenBank mRNA | BC103292.1 | - | 2 - 3 | 372 | 159 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
