Assay Details
Gene Symbol: PLVAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035353.2 | NP_001030430.1 | 2 - 3 | 514 | 79 | |
GenBank mRNA | BC102834.1 | - | 2 - 3 | 517 | 79 | |
BT025486.1 | - | 2 - 3 | 465 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
