Assay Details
Gene Symbol: F3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173878.2 | NP_776303.1 | 4 - 5 | 736 | 140 | |
GenBank mRNA | BC123808.1 | - | 4 - 5 | 709 | 140 | |
S74147.1 | - | 4 - 5 | 736 | 140 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
