Assay Details
Gene Symbol: NPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173918.2 | NP_776343.1 | 2 - 3 | 224 | 73 | |
GenBank mRNA | BC102504.1 | - | 2 - 3 | 212 | 73 | |
BT021533.1 | - | 2 - 3 | 203 | 73 | ||
X85799.1 | - | 224 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
