Assay Details
Gene Symbol: NPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173918.2 | NP_776343.1 | 3 - 4 | 401 | 95 | |
GenBank mRNA | BT021533.1 | - | 3 - 4 | 380 | 95 | |
X85799.1 | - | 401 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
