Assay Details
Gene Symbol: IL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173920.2 | NP_776345.1 | 1 - 2 | 177 | 124 | |
GenBank mRNA | L31893.1 | - | 1 - 2 | 177 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
