Assay Details
Gene Symbol: CXCL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173925.2 | NP_776350.1 | 2 - 3 | 265 | 105 | |
GenBank mRNA | AF232704.1 | - | 2 - 3 | 265 | 105 | |
BC103310.1 | - | 2 - 3 | 254 | 105 | ||
EU276073.1 | - | 2 - 3 | 236 | 105 | ||
JN559767.1 | - | 2 - 3 | 236 | 105 | ||
S82598.1 | - | 2 - 3 | 192 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
