Assay Details
Gene Symbol: MIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173937.2 | NP_776362.1 | 3 - 4 | 654 | 79 | |
GenBank mRNA | K02818.1 | - | 3 - 4 | 654 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
