Assay Details
Gene Symbol: MX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173940.2 | NP_776365.1 | 12 - 13 | 1575 | 126 | |
GenBank mRNA | AB060169.1 | - | 11 - 12 | 1951 | 126 | |
AF047692.1 | - | 11 - 12 | 1497 | 126 | ||
BC118109.1 | - | 12 - 13 | 1564 | 126 | ||
JQ766265.1 | - | 11 - 12 | 1477 | 126 | ||
U88329.1 | - | 12 - 13 | 1575 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
