Assay Details
Gene Symbol: CTSV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174032.2 | NP_776457.1 | 7 - 8 | 992 | 89 | |
GenBank mRNA | BC102312.1 | - | 7 - 8 | 1038 | 89 | |
BC151425.1 | - | 7 - 8 | 992 | 89 | ||
BT021022.1 | - | 7 - 8 | 968 | 89 | ||
X91755.1 | - | 992 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
