Assay Details
Gene Symbol: DSG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174045.1 | NP_776470.1 | 14 - 15 | 2216 | 81 | |
GenBank mRNA | X57784.1 | - | 1987 | 81 | ||
X58466.1 | - | 2216 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
