Assay Details
Gene Symbol: FMOD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174058.2 | NP_776483.1 | 1 - 2 | 58 | 71 | |
GenBank mRNA | BC104615.1 | - | 1 - 2 | 56 | 71 | |
X16485.1 | - | 58 | 71 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC104615.1, NM_174058.2, X16485.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
