Assay Details
Gene Symbol: POMC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174151.1 | NP_776576.1 | 1 - 2 | 95 | 127 | |
GenBank mRNA | M25587.1 | - | 1 - 2 | 95 | 127 | |
V00107.1 | - | 95 | 127 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_174151.1, V00107.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
