Assay Details
Gene Symbol: PPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174154.2 | NP_776579.1 | 8 - 9 | 810 | 62 | |
GenBank mRNA | BC134479.1 | - | 8 - 9 | 808 | 62 | |
L34261.1 | - | 8 - 9 | 810 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
