Assay Details
Gene Symbol: GRK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174173.2 | NP_776598.1 | 3 - 4 | 1181 | 65 | |
GenBank mRNA | M73836.1 | - | 3 - 4 | 1181 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
