Assay Details
Gene Symbol: GRK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174173.2 | NP_776598.1 | 6 - 7 | 1595 | 139 | |
GenBank mRNA | M73836.1 | - | 6 - 7 | 1595 | 139 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
