Assay Details
Gene Symbol: SARS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174175.3 | NP_776600.1 | 10 - 11 | 1473 | 73 | |
GenBank mRNA | AF297553.1 | - | 10 - 11 | 1405 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
