Assay Details
Gene Symbol: WARS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174218.1 | NP_776643.1 | 9 - 10 | 1440 | 95 | |
GenBank mRNA | BC102806.1 | - | 10 - 11 | 1388 | 95 | |
X52113.1 | - | 1367 | 95 | |||
X53918.1 | - | 1440 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
