Assay Details
Gene Symbol: ADRB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174232.2 | NP_776657.1 | 1 - 2 | 1298 | 75 | |
GenBank mRNA | BC116092.1 | - | 1 - 2 | 1297 | 75 | |
X85961.1 | - | 1298 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
