Assay Details
Gene Symbol: HAPLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174288.1 | NP_776713.1 | 2 - 3 | 208 | 120 | |
GenBank mRNA | BC151455.1 | - | 2 - 3 | 335 | 120 | |
U02292.1 | - | 2 - 3 | 208 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
