Assay Details
Gene Symbol: GLRA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174321.2 | NP_776746.1 | 7 - 8 | 1137 | 58 | |
GenBank mRNA | AF268375.1 | - | 7 - 8 | 1137 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
