Assay Details
Gene Symbol: NR5A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174403.2 | NP_776828.1 | 2 - 3 | 224 | 76 | |
GenBank mRNA | D13569.1 | - | 2 - 3 | 224 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
