Assay Details
Gene Symbol: RPE65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174453.2 | NP_776878.1 | 1 - 2 | 63 | 93 | |
GenBank mRNA | L11356.1 | - | 1 - 2 | 63 | 93 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_174453.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
