Assay Details
Gene Symbol: RPE65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174453.2 | NP_776878.1 | 9 - 10 | 1047 | 92 | |
GenBank mRNA | L11356.1 | - | 9 - 10 | 1047 | 92 | |
X66277.1 | - | 1001 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
