Assay Details
Gene Symbol: IGFBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174555.1 | NP_776980.1 | 2 - 3 | 649 | 93 | |
GenBank mRNA | AF074854.1 | - | 2 - 3 | 649 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
