Assay Details
Gene Symbol: PPP3R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174583.2 | NP_777008.1 | 3 - 4 | 281 | 96 | |
GenBank mRNA | AB375794.1 | - | 1 - 2 | 170 | 96 | |
X71666.1 | - | 281 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
