Assay Details
Gene Symbol: PPP3R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174583.2 | NP_777008.1 | 5 - 6 | 526 | 133 | |
GenBank mRNA | X71666.1 | - | 526 | 133 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
