Assay Details
Gene Symbol: CD40LG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174624.2 | 2 - 3 | 297 | 141 | ||
GenBank mRNA | Z48469.1 | - | 297 | 141 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
