Assay Details
Gene Symbol: PRDX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174643.1 | NP_777068.1 | 4 - 5 | 589 | 65 | |
GenBank mRNA | AF080228.1 | - | 4 - 5 | 613 | 65 | |
AF090194.1 | - | 4 - 5 | 589 | 65 | ||
AJ243848.1 | - | 4 - 5 | 599 | 65 | ||
BC102172.1 | - | 4 - 5 | 614 | 65 | ||
BT020967.1 | - | 4 - 5 | 603 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
