Assay Details
Gene Symbol: GABARAPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174675.2 | NP_777100.1 | 2 - 3 | 169 | 99 | |
GenBank mRNA | AF020262.1 | - | 2 - 3 | 169 | 99 | |
AY117147.1 | - | 2 - 3 | 129 | 99 | ||
BC102902.1 | - | 2 - 3 | 190 | 99 | ||
BT020958.1 | - | 2 - 3 | 178 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
