Assay Details
Gene Symbol: MGP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174707.3 | NP_777132.1 | 1 - 2 | 93 | 86 | |
GenBank mRNA | AB098895.1 | - | 1 - 2 | 106 | 86 | |
AF210379.1 | - | 1 - 2 | 93 | 86 | ||
AY911361.1 | - | 1 - 2 | 117 | 86 | ||
BC102587.1 | - | 1 - 2 | 123 | 86 | ||
X07363.1 | - | 64 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
