Assay Details
Gene Symbol: CHRNA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174719.2 | NP_777144.1 | 1 - 2 | 412 | 150 | |
GenBank mRNA | X57032.1 | - | 412 | 150 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_174719.2, X57032.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
