Assay Details
Gene Symbol: CHRNA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174719.2 | NP_777144.1 | 2 - 3 | 552 | 74 | |
GenBank mRNA | BC142063.1 | - | 2 - 3 | 229 | 74 | |
X57032.1 | - | 552 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
