Assay Details
Gene Symbol: RPS27A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174778.1 | NP_777203.1 | 2 - 3 | 101 | 114 | |
GenBank mRNA | AB098891.1 | - | 2 - 3 | 228 | 114 | |
AF058700.1 | - | 2 - 3 | 101 | 114 | ||
BC102491.1 | - | 3 - 4 | 178 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
