Assay Details
Gene Symbol: RPS27A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174778.1 | NP_777203.1 | 4 - 5 | 319 | 93 | |
GenBank mRNA | AB098891.1 | - | 4 - 5 | 446 | 93 | |
AF058700.1 | - | 4 - 5 | 319 | 93 | ||
BC102491.1 | - | 5 - 6 | 396 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
