Assay Details
Gene Symbol: GUCA1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174786.1 | NP_777211.1 | 3 - 4 | 579 | 80 | |
GenBank mRNA | L43001.1 | - | 3 - 4 | 579 | 80 | |
U32856.1 | - | 3 - 4 | 480 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
