Assay Details
Gene Symbol: TSPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175776.2 | NP_786970.1 | 1 - 2 | 39 | 104 | |
GenBank mRNA | BC102933.1 | - | 1 - 2 | 39 | 104 | |
M64520.1 | - | 1 - 2 | 39 | 104 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC102933.1, NM_175776.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
