Assay Details
Gene Symbol: TSPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175776.2 | NP_786970.1 | 2 - 3 | 240 | 83 | |
GenBank mRNA | BC102933.1 | - | 2 - 3 | 240 | 83 | |
M64520.1 | - | 2 - 3 | 240 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
