Assay Details
Gene Symbol: GUCY1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175778.2 | NP_786972.1 | 3 - 4 | 610 | 95 | |
GenBank mRNA | KX258202.1 | - | 3 - 4 | 610 | 95 | |
X54014.1 | - | 610 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
