Assay Details
Gene Symbol: NDUFB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175787.2 | NP_786981.1 | 1 - 2 | 268 | 158 | |
GenBank mRNA | BC102533.1 | - | 1 - 2 | 232 | 158 | |
X63212.1 | - | 268 | 158 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
