Assay Details
Gene Symbol: GLTP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175799.3 | NP_786993.1 | 1 - 2 | 110 | 137 | |
GenBank mRNA | AF209701.1 | - | 1 - 2 | 111 | 137 | |
BC105256.1 | - | 1 - 2 | 216 | 137 | ||
BC142445.1 | - | 1 - 2 | 124 | 137 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
