Assay Details
Gene Symbol: GLTP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175799.3 | NP_786993.1 | 3 - 4 | 308 | 146 | |
GenBank mRNA | AF209701.1 | - | 3 - 4 | 309 | 146 | |
BC105256.1 | - | 3 - 4 | 414 | 146 | ||
BC142445.1 | - | 3 - 4 | 322 | 146 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
