Assay Details
Gene Symbol: CCL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175827.2 | NP_787021.1 | 2 - 3 | 237 | 117 | |
GenBank mRNA | AJ007043.2 | - | 2 - 3 | 237 | 117 | |
BC102064.1 | - | 2 - 3 | 244 | 117 | ||
EU276060.1 | - | 2 - 3 | 185 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
