Assay Details
Gene Symbol: HMGB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176612.1 | NP_788785.1 | 4 - 5 | 595 | 97 | |
GenBank mRNA | BC102929.1 | - | 4 - 5 | 581 | 97 | |
BT030587.1 | - | 4 - 5 | 616 | 97 | ||
M26110.1 | - | 1 - 2 | 132 | 97 | ||
X12796.1 | - | 595 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
